Learn More
Merck Montage™ SEQ384 Sequencing Reaction Cleanup Kit
Product Code. 10721073
Click to view available options
Quantity:
10 Pack
50 Pack
Unit Size:
Pack of 10
Pack of 50
Product Code. 10721073
Brand: Merck S384SEQ50
Requires
MultiScreen™ 384 Vacuum Manifold with underdrain support grid (Mfr. No. MAVM 096 0R)
Use 384-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
  • Recovered sequencing products are highly purified and result in long, accurate sequencing reads
  • Protocols are optimized for use with BigDye™ Terminator version 3.0, 3.1, 1.1 and ET Terminator sequencing chemistries
  • Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
  • Plates are suitable for use with 1/8 sequencing reactions, reducing reagent costs by miniaturization
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) DNA sequencing
Includes Clear Polystyrene 384-well plates
Quantity 50 Pack