missing translation for 'onlineSavingsMsg'
Learn More

Merck Millistak+™ HC Pod Disposable Depth Filter System

Product Code. 15822305 Shop All Merck Products
Change view
Click to view available options
Grade:
C0HC
Filtration Area:
0.027 m2
For Use With (Application):
Perfusion bioreactor fluid, laboratory filtration
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grade Filtration Area For Use With (Application) unitSize
15822305 C0HC 0.027 m2 Perfusion bioreactor fluid, laboratory filtration 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15822305 Supplier Merck Supplier No. MC0HC027H1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Combine two distinct technologies to enhance filter capacity and retention

  • Filter material: cellulose and inorganic filter aid
  • Configurations: 0.11, 0.55, or 1.1 m2 filtration area
  • Low hold-up volume for greater product yield
  • Broad range of 23 media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000L or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Disposable Pod system protects operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Ideal for Primary and Secondary Clarification in Lab, Pilot and Process-Scale Applications

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material Glass Fiber-reinforced Polypropylene
Material (Membrane) Cellulose
Filtration Area 0.027 m2
For Use With (Application) Perfusion bioreactor fluid, laboratory filtration
Grade C0HC
Inlet Connection (English) 0.25 in. hose
Max. Pressure 50 psi at 25°C, 15 psi at 80°C
No. per Pack 1 Ea.
Type Millistak+ Lab Scale Pod Filter
Length (English) 8.5 in.
Length (Metric) 22 cm
Height (English) 5.3 in.
Height (Metric) 14 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.