missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMD Millipore™ Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophobic PTFE
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Brand: EMD Millipore™ SLFH025NS
Code : 71
Additional Details : Weight : 0.10000kg
Description
- Compatible with more aggressive organic solutions and high temperature conditions
Specifications
Syringe Filter | |
Hydrophobic Polytetrafluoroethylene (PTFE) | |
0.45μm | |
50/Pk. |
25mm | |
Polyethylene | |
No |