Learn More
Merck™ Microfil™ V Filtration Device
Filter water and other beverages.  Microfil™ V Filtration Device is a sterile filter that can contain 100mL.
£190.00 - £206.00
Specifications
Description Fitration Device
View More Specs
Includes: Filtration device; 100mL polypropylene funnel; white, gridded sterile 47mm MCE membrane with 0.22 or 0.45μm pore size.
Products 2
Product Code Brand Material Pore Size Price Quantity & Availability  
Product Code Brand Material Pore Size Price Quantity & Availability  
10571078 View Documents Merck™
MVGSWG124
MCE, White, gridded 0.22μm £206.00
Pack of 24 View alternative products
Save Up to 
»
Estimated Shipment
24-02-2020
Add to basket
13118013 View Documents Merck™
MVHAWG124
MCE, White, gridded 0.45μm £190.00
Pack of 24 View alternative products
Save Up to 
»
In Stock 28
Add to basket
Description
Description
Uniform microorganism recovery is promoted by unique “push-fit” seal; smooth, hydrophobic funnel surface reduces aqueous sample residues sticking to funnel walls; and support's convex shape eliminates membrane distortion, thus maintaining uniform contact between membrane and medium surface
  • No need to assemble filter holders, clean funnels, or sterilize filter support after each filtration
  • Microfil V filtration device snaps off the carrying tray and onto a standard No. 8 stopper or a vacuum syringe with Luer connection
  • Funnels attach with “push-fit” seal to prevent sample bypass without clamps or O-rings
  • Filter support has a curved edge to easily remove membrane after filtration while vacuum is still applied
Order Info
Sold in three trays of eight
Certifications
Membrane is certified in accordance with Standard Methods for Coliform Analysis.
Compliance
Conforms to international standards for testing drinking water, Standard Methods for the Examination of Water and Wastewater, and WHO Guidelines for Drinking Water Quality.
Specifications
Specifications
Fitration Device