Learn More
Merck MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free
Composed of mixed cellulose esters (nitrate and acetate)
Brand: Merck HATF04700
Code : 71
Additional Details : CAS Number : 9004-70-0 Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
Composed of mixed cellulose esters (nitrate and acetate)
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.Features & Benefits: - Versatile filter for biological and environmental monitoring applications - Available in a range of pore sizes, colored black or white, with or without a gridded surface - Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methodsSpecifications
Not Autoclavable | |
Membrane Filter | |
PVDF | |
Non-sterile | |
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.). | |
Blotting Membranes | |
47 mm |
White | |
Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free | |
0.45 μm | |
60 mL/min x cm2 | |
Plain | |
Hydrophilic | |
47 mm |
Safety and Handling
- IMMO-NC Triton Free MCE .45um Wh PL 100pk
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.