missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MF-Millipore™ Membrane Filter, 5 μm
Mixed cellulose esters (MCE) membrane, hydrophilic
Brand: Merck SMWP14250
This item is not returnable.
View return policy
Description
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
Not Autoclavable | |
White | |
5 μm | |
580 mL/min x cm2 | |
Hydrophilic | |
50Pack | |
135 μm | |
Hydrophilic | |
142 mm |
≥0.42 bar, air with water | |
MCE | |
Non-sterile | |
32 L/min/ cm2 | |
Plain | |
50/Pk. | |
Filter Membranes | |
142 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- MCE 5.0UM WH PL 142MM 50/PK
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use to extinguish.