missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Luminata™ Classico Western HRP substrate, 100mL
Shop All Merck Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Beskrivning
- The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays.
- Premixed for convenience to avoid cross contamination of the reagents.
- Broad range of sensitivities to satisfy all detection needs.
- Consistent results with less pipetting error.
- Stable at 4°C and also at room temperature which allows storage on bench.
- Best detection performance relative to competition.
- Compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Western blotting, dot/slot blotting
Specifikationer
Specifikationer
| Quantity | 100 mL |
| Storage Requirements | 2° to 8°C |
| For Use With (Application) | Dot/Slot Blotting, Western Blotting |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering