missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immobilon-PSQ Membrane:0.2 μm pore size and a large internal structure Higher protein adsorption and sequencing yields than other membranes Recommended for blotting proteins less than 20 kDa Prevents blow-through of low molecular weight proteins
Specifications
Specifications
| Length (Metric) | 7 cm |
| Width (Metric) | 8.4 cm |
| For Use With (Application) | For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins |
| Color | White |
| Code | ISEQ |
| Surface | Plain |
| Width (English) | 3.30 in. |
| Format | Sheet |
| Material | Immobilon-PSQ |
| Length (English) | 2.75 in. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?