missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™-PSQ PVDF Membranes

Product Code. 11506345 Shop All Merck Products
Click to view available options
Dimensions (LxW):
10 m x 8.5 cm
10 x 10 cm
15 x 15 cm
20 x 20 cm
26 x 26 cm
26.5 cm x 3.75 m
7 x 8.4 cm
8 x 10 cm
8.5 x 13.5 cm
9 x 12 cm
Format:
Roll
Sheet
Unit Size:
1 roll
Each
Pack of 10
Pack of 50
This item is not returnable. View return policy

Product Code. 11506345

Brand: Merck ISEQ08100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Western blots and protein sequencing

Immobilon-PSQ Membrane:
  • 0.2 μm pore size and a large internal structure
  • Higher protein adsorption and sequencing yields than other membranes
  • Recommended for blotting proteins less than 20 kDa
  • Prevents blow-through of low molecular weight proteins
  • TRUSTED_SUSTAINABILITY

    Specifications

    Length (Metric) 8 cm
    Width (Metric) 10 cm
    For Use With (Application) For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins
    Format Sheet
    Material Immobilon-PSQ
    Pore Size 0.2 μm
    Dimensions (LxW) 8 x 10 cm
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.