missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Immobilon™-PSQ PVDF Membranes
For Western blots and protein sequencing
Brand: Merck ISEQ26260
458.45 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Immobilon-PSQ Membrane:Specifications
26 cm | |
For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins | |
Immobilon-PSQ | |
26 x 26 cm |
26 cm | |
Sheet | |
0.2 μm |