Learn More
Merck Adapter, 1/4 in. NPTF to F Luer-Lok
Accessories for filter holders for sample preparation.
Brand:  Merck XX3002568
Product Code. 10345381
  • ONLINE EXCLUSIVE PRICE
    £177.00
    £162.11 / Pack of 1
    Save £14.89
    8% Off
Only null left
Estimated Shipment: 06-07-2026
Add to basket
Description
Description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.