missing translation for 'onlineSavingsMsg'
Learn More

Merck Express™ PLUS Membrane Filters

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

Brand:  Merck GPWP04700

204.47 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 71

Additional Details : Weight : 0.14130kg

 View more versions of this product

Product Code. 10228250

  • £363.00 / Pack of 100
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.Features & Benefit:•Ultra fast filtration and high throughput shorten process time •Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specifications

Specifications

Not Autoclavable
Screen Filter
PES
Non-sterile
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Plain
Hydrophilic
47 mm
White
PES
0.22 μm
15 (at 690 Mbar (10 psi))
Hydrophilic
Filter Membranes
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers