missing translation for 'onlineSavingsMsg'
Learn More

Merck Chemicon™ EndoGRO™-VEGF Complete Culture Media Kit

Product Code. 17704909 Shop All Merck Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17704909 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17704909 Supplier Merck Supplier No. SCME002
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

a low serum formulation that contains VEGF, It is intended for rapid proliferation of Human Endothelial Cells, HUVEC, aortic endothelial cells and other large vessel endothelial cells.

EndoGRO Media Formulations provide an optimal cell culture environment for many types of endothelial cells, including HUVEC, aortic endothelial cells and other human large vessel and microvascular endothelial cells. These media have been shown to grow endothelial cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge that helps prevent contamination. EndoGRO Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results. Four different media formulations are available for various applications and cell culture requirements.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell culture
Quantity 500 mL
Content And Storage Basal medium: 2-8°C in dark; Supplement Mix: -20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.