missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ready-to-use
- Protein-free, reducing nonspecific binding, especially when using phosphoantibodies or avidin-biotin systems
- Specially engineered for the EMD Millipore SNAP i.d.™ detection system, but may be used in traditional Western blotting protocols
- Suitable for the dilution of primary and secondary antibodies
- Sufficient for 19 miniblots (each 8 × 7cm blot requires at least 26mL for SNAP i.d.)
- Compatible with streptavidin antibodies
- Compatible with PVDF (Immobilon™ P, Immobilon FL) and nitrocellulose
- Block-CH reagent is compatible with Immobilon HRP substrate
- Choose from three Block reagents
- Block-CH reagent for HRP chemiluminescence detection
- Block-FL reagent for fluorescence detection
- Block-PO reagent for phosphoprotein detection via chemiluminescence or fluorescence
Specifications
Specifications
| Product Type | Block-PO Reagent |
| For Use With (Application) | Mass Spectrometry |
| Quantity | 500 mL |
| Sufficient For | Nineteen 8 x 7cm miniblots |
| Content And Storage | 20°C to 25°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?