missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Pro Purification System with Ultra-0.5 Device, 3 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Brand: Merck ACS500312
Code : NEW
Additional Details : Weight : 0.50000kg
Description
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Non-sterile | |
12 Amicon Ultra NMWCO 3 kDa filters, 12 Exchange devices, and 12 Collection tubes with cap | |
Centrifugal Filters | |
12 cm |
3 KDa | |
Regenerated Cellulose (RC) | |
12Pack | |
15 mL |