Learn More
Merck Amicon™ Pro Affinity Concentration Kit Ni-NTA with 3kDa Amicon™ Ultra-0.5 Device
Amicon Pro Affinity Concentration Kit Ni-NTA - A Centrifugal Tool for purifying and concentrating His-tagged proteins.
Brand: Merck ACK5003NT
Code : NEW
Additional Details : CAS Number : 64-17-5 Weight : 3.00000kg
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3 ml HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25ml Bind Buffer (4X), 25 ml Wash buffer (4X), 5ml elution buffer (4X)Specifications
Clear with pink cap | |
Ultracel Low-binding Regenerated Cellulose | |
Non-sterile | |
Antibody labeling, Protein purification, Protein concentration, Buffer exchange, Protein enrichment | |
12Pack Amicon Pro, 12Pack Amicon Ultra NMWCO 3 kd, 3 mL HisBind Ni-NTA resin (Binding capacity: 5 to 10 mg His tagged protein/ mL agarose), 25 mL Bind Buffer (4 x), 25 mL Wash buffer (4 x), 5 mL elution buffer (4 x) | |
12Pack | |
10 mL | |
12 cm | |
29.7 mm |
Single-use | |
3 kDa | |
1 cm2 | |
Regenerated Cellulose (RC) | |
4000 xg for concentration and buffer exchange spin; 1000 xg for flow through, wash, elution and reverse spin | |
Centrifugal Filters | |
4.7 in. | |
1.2 in. |