Learn More
Merck Accessories for XX2702550 1225 Sampling Manifold
Product Code. 10693202 Shop All Merck Products
Click to view available options
Type:
Ball Valve
Chamber Replacement Kit
O-Ring
Sample Cup Extension Barrel
Support Plate
Unit Size:
Each
Pack of 12
Pack of 30
Product Code. 10693202
Brand: Merck XX11000PP
Only null left
Add to basket
Add to basket
Accessories for filter holders for sample preparation.
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Connectors 1/4 in. Stepped hose connector / 1/4 in. NPTM
For Use With (Equipment) 1225 sampling manifold
Material Glass-filled Polypropylene
Type Ball Valve