missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, 0.1 μm

Product Code. 10024450 Shop All Merck Products
Click to view available options
Diameter (Metric):
25 mm
47 mm
90 mm
105 mm
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 10024450

Brand: Merck VCWP04700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features & Benefits:

  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color White
Material MCE
Pore Size 0.1 μm
Sterility Non-sterile
Flow Rate 1.5 mL/min x cm2
Thickness (Metric) 105 μm
Type Filter Membranes
Wettability Hydrophilic
Diameter (Metric) 47 mm
compliance-icons
Product Identifier
  • MCE 0.1UM WH PL 47MM 100/PK
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.