missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EMD 386088 hydrochloride
GREENER_CHOICE

Product Code. 18094842
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094842 50 mg 50mg
18726026 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094842 Supplier Tocris Bioscience™ Supplier No. 2382/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent 5-HT6 agonist

EMD 386088 hydrochloride is a potent 5-HT6 receptor agonist (EC50 = 1.0 nM) that displays selectivity over other 5-HT receptors (IC50 values are 7.4, 110, 180, 240, 450, 620, 660 and 3000 nM for 5-HT6, 5-HT1D, 5-HT1B, 5-HT2A, 5-HT2C, 5-HT4, 5-HT1A and 5-HT7 receptors respectively). Shows moderate affinity at 5-HT3 receptors (IC50 = 34 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole hydrochloride
CAS 1171123-46-8
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C14H15N2Cl.HCl
Formula Weight Observed MW: 283.2

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.