missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EMD 386088 hydrochloride

Description
EMD 386088 hydrochloride is a potent 5-HT6 receptor agonist (EC50 = 1.0 nM) that displays selectivity over other 5-HT receptors (IC50 values are 7.4, 110, 180, 240, 450, 620, 660 and 3000 nM for 5-HT6, 5-HT1D, 5-HT1B, 5-HT2A, 5-HT2C, 5-HT4, 5-HT1A and 5-HT7 receptors respectively). Shows moderate affinity at 5-HT3 receptors (IC50 = 34 nM).
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-2-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole hydrochloride |
| CAS | 1171123-46-8 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C14H15N2Cl.HCl |
| Formula Weight | Observed MW: 283.2 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?