missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMC4 Recombinant Rabbit Monoclonal Antibody (K01_2J38)
Description
Immunogen sequence: LYPVGYLDKQ VPDT.
May mediate anti-apoptotic activity.
Specifications
Specifications
| Antigen | EMC4 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2J38 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | EMC4 |
| Gene Accession No. | Q5J8M3, Q9CZX9 |
| Gene Alias | 2610318K02Rik; Cell proliferation-inducing gene 17 protein; EMC4; ER membrane protein complex subunit 4; HSPC184; PIG17; TMEM85; Transmembrane protein 85 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?