Learn More
Cole-Parmer™ Embosser Tape for Manual Laboratory Label Maker
Product Code. 17317639 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17317639 Blue Each
17397629 Black Each
17307639 Red Each
17327639 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17317639 Supplier Cole-Parmer™ Supplier No. 6554348
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Embosser Tape for the Manual Laboratory Label Maker sticks firmly to glass, plastic, metal, rubber, and wood.
  • Easily remove back adhesive shield
  • Choose from four colors to color-code your lab
Specifications
Product Type Colored Label Tape
Color Blue
Length (English) 118 in.
Width (English) 3/8 in.
Length (Metric) 300 cm
Width (Metric) 0.95 cm
Dimensions (L x W) 118 x 3/8 in.
Adhesive Type Peel and Stick
Sterility Non-sterile
Color Capability Monochrome
Format Label