missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Embigin homolog Polyclonal specifically detects Embigin homolog in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Embigin homolog |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | embigin, embigin homolog, embigin homolog (mouse), F9, H, MGC71745 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 33-260 of human Embigin homolog (NP_940851.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?