missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Embelin, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Embelin |
| CAS | 550-24-3 |
| Molecular Formula | C17 H26 O4 |
| Quantity | 10 mg |
| Synonym | 2,5-Dihydroxy-3-undecyl-p-benzoquinone, 2,5-Dihydroxy-3-undecyl-1,4-benzoquinone, Embelic acid, Emberine, NSC 91874, 2,5-Dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione |
| InChI Formula | InChI=1S/C17H26O4/c1-2-3-4-5-6-7-8-9-10-11-13-16(20)14(18)12-15(19)17(13)21/h12,18,21H,2-11H2,1H3 |
| SMILES | CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O |
| IUPAC Name | 2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 294.39 |
| Formula Weight | 294.1831 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction