missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Tissue Embedding Cassettes, Mega Type

Product Code. 18038021
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
18038021 Blue Pack of 1000
18068041 Yellow Pack of 1000
18098021 Green Pack of 1000
18078041 Pink Pack of 1000
10053011 White Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18038021 Supplier Kartell™ Supplier No. 0292504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tissue embedding cassettes, mega type, for medium to large-sized samples.

  • Cassette for laboratory use
  • To be used for paraffin inclusions
  • Mega type
  • For inclusion and treatment of human samples and to perform laboratory tests
  • CE marked, IVD certified
  • Clipon closure lid that can easily be removed

Specifications

Quantity 1000 Pk.
Color Blue
Product Type Embedding Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.