missing translation for 'onlineSavingsMsg'
Learn More

Emapunil, MedChemExpress

Product Code. 16492687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492687 50 mg 50mg
16466598 10 mg 10mg
16459167 5 mg 5mg
16426796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492687 Supplier Medchem Express Supplier No. HY15527
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Emapunil (AC-5216), an orally active and selective TSPO (a mitochondrial benzodiazepine receptor) ligand, produces anti-anxiety and antidepressant-like effects in various animal models.

Emapunil (AC-5216), an orally active and selective TSPO (a mitochondrial benzodiazepine receptor) ligand, produces anti-anxiety and antidepressant-like effects in various animal models.

Specifications

Chemical Name or Material Emapunil
CAS 226954-04-7
Color White
Purity Grade Notes Research
Molecular Formula C23H23N5O2
Quantity 50 mg
Synonym AC-5216 XBD-173
Solubility Information DMSO : 33.33 mg/mL (83.02 mM; Need ultrasonic)
SMILES O=C(N(CC)CC1=CC=CC=C1)CN2C(N(C)C3=CN=C(C4=CC=CC=C4)N=C23)=O
Molecular Weight (g/mol) 401.46
Formula Weight 401.46
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.