Learn More
Invitrogen™ Emapalumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559497
 View more versions of this product
Product Code. 30284618
  • £397.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Emapalumab, also known as NI-0501, is a fully human monoclonal antibody that targets interferon gamma. Emapalumab development was sponsored by NovImmune SA, further developed by Sobi and FDA approved on November 20, 2018. 1, 5 The approval of emapalumab was followed by the designation of orphan pharmaceutical, priority review and breakthrough therapy. 5 As well, emapalumab was given the status of PRIME by the EMA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Emapalumab
Recombinant Monoclonal
Unconjugated
emapalumab-lzsg; NI-0501
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized