missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMAP-II/AIMP1 Antibody (S01-5A9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19624-50ul
This item is not returnable.
View return policy
Description
EMAP-II/AIMP1 Monoclonal antibody specifically detects EMAP-II/AIMP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| EMAP-II/AIMP1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-5A9 | |
| Unconjugated | |
| aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43 | |
| A synthetic peptide of human EMAP-II/AIMP1 (Uniprot # Q12904) | |
| 50 μg | |
| Biologically Active Proteins, Cytokine Research | |
| 9255 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction