missing translation for 'onlineSavingsMsg'
Learn More

EMAP-II/AIMP1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18351724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351724 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351724 Supplier Novus Biologicals Supplier No. NBP299128FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EMAP-II/AIMP1 Polyclonal specifically detects EMAP-II/AIMP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMAP-II/AIMP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant EMAP-II/AIMP1 (NP_004748.2; Ala2-Lys312).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9255
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.