missing translation for 'onlineSavingsMsg'
Learn More

Emamectin Benzoate (90%), TRC

Product Code. 30040514
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040514 100 mg 100mg
30009240 1 g 1g
30003793 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040514 Supplier Toronto Research Chemicals Supplier No. TRCE520500100MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155569-91-8
Molecular Formula C49 H75 N O13 . C48 H73 N O13 . 2 C7 H6 O2
Molecular Weight (g/mol) 2002.45
Synonym Avermectin B1, 4' '-deoxy-4' '-(methylamino)-, (4' 'R)-, benzoate (1:1), Avermectin B1, 4' '-deoxy-4' '-(methylamino)-, (4' 'R)-, benzoate (salt) (9CI), (4' 'R)-4' '-Deoxy-4' '-(methylamino)avermectin B1 benzoate, Affirm, Denim, Denim (pesticide), Emamectin benzoate, Emamectin benzoate (salt), MK 244, Methylamino abamectin benzoate, Proclaim, Proclaim 05SG, Proclaim 5SG, Revive, Revive (insecticide), Sch 58854, Shot Wan Liquid Formulation, Shot-Wan, Slice, Tree-age
SMILES CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@H](NC)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C.CN[C@@H]8[C@H](C)O[C@H](C[C@@H]8OC)O[C@H]9[C@H](C)O[C@H](C[C@@H]9OC)O[C@H]%10[C@@H](C)\C=C\C=C\%11/CO[C@@H]%12[C@H](O)C(=C[C@@H](C(=O)O[C@H]%13C[C@@H](C\C=C\%10/C)O[C@@]%14(C%13)O[C@H](C(C)C)[C@@H](C)C=C%14)[C@]%11%12O)C.OC(=O)c%15ccccc%15.OC(=O)c%16ccccc%16

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C49H75NO13.C48H73NO13.2C7H6O2/c1-12-26(2)44-29(5)18-19-48(63-44)24-35-21-34(62-48)17-16-28(4)43(27(3)14-13-15-33-25-56-46-42(51)30(6)20-36(47(52)59-35)49(33,46)53)60-40-23-38(55-11)45(32(8)58-40)61-39-22-37(54-10)41(50-9)31(7)57-39;1-25(2)42-28(5)17-18-47(62-42)23-34-20-33(61-47)16-15-27(4)43(26(3)13-12-14-32-24-55-45-41(50)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-11)44(31(8)57-39)60-38-21-36(53-10)40(49-9)30(7)56-38;2*8-7(9)6-4-2-1-3-5-6/h13-16,18-20,26-27,29,31-32,34-46,50-51,53H,12,17,21-25H2,1-11H3;12-15,17-19,25-26,28,30-31,33-45,49-50,52H,16,20-24H2,1-11H3;2*1-5H,(H,8,9)/b14-13+,28-16+,33-15+;13-12+,27-15+,32-14+;;/t26-,27-,29-,31-,32-,34+,35-,36-,37-,38-,39-,40-,41+,42+,43-,44+,45-,46+,48+,49+;26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41+,42+,43-,44-,45+,47+,48+;;/m00../s1
Formula Weight 2001.1056
Chemical Name or Material Emamectin benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.