missing translation for 'onlineSavingsMsg'
Learn More

Emamectin benzoate 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30328061
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328061 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328061 Supplier Dr. Ehrenstorfer Supplier No. DREA13117000AL50
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Emamectin benzoate
CAS 155569-91-8
Analyte or Component Names Emamectin benzoate
Shipping Condition +20°C
Molecular Formula C49 H75 N O13 . C48 H73 N O13 . 2 C7 H6 O2
Concentration 50 μg/mL
Certifications/Compliance ISO 17034
Synonym Avermectin B1, 4''-deoxy-4''-(methylamino)-, (4''R)-, benzoate (1:1), Avermectin B1, 4''-deoxy-4''-(methylamino)-, (4''R)-, benzoate (salt) (9CI), (4''R)-4''-Deoxy-4''-(methylamino)avermectin B1 benzoate, Affirm, Denim, Denim (pesticide), Emamectin benzoate, Emamectin benzoate (salt), MK 244, Methylamino abamectin benzoate, Proclaim, Proclaim 05SG, Proclaim 5SG, Revive, Revive (insecticide), Sch 58854, Shot Wan Liquid Formulation, Shot-Wan, Slice, Tree-age
InChI Formula InChI=1S/C49H75NO13.C48H73NO13.2C7H6O2/c1-12-26(2)44-29(5)18-19-48(63-44)24-35-21-34(62-48)17-16-28(4)43(27(3)14-13-15-33-25-56-46-42(51)30(6)20-36(47(52)59-35)49(33,46)53)60-40-23-38(55-11)45(32(8)58-40)61-39-22-37(54-10)41(50-9)31(7)57-39;1-25(2)42-28(5)17-18-47(62-42)23-34-20-33(61-47)16-15-27(4)43(26(3)13-12-14-32-24-55-45-41(50)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-11)44(31(8)57-39)60-38-21-36(53-10)40(49-9)30(7)56-38;2*8-7(9)6-4-2-1-3-5-6/h13-16,18-20,26-27,29,31-32,34-46,50-51,53H,12,17,21-25H2,1-11H3;12-15,17-19,25-26,28,30-31,33-45,49-50,52H,16,20-24H2,1-11H3;2*1-5H,(H,8,9)/b14-13+,28-16+,33-15+;13-12+,27-15+,32-14+;;/t26-,27-,29-,31-,32-,34+,35-,36-,37-,38-,39-,40-,41+,42+,43-,44+,45-,46+,48+,49+;26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41+,42+,43-,44-,45+,47+,48+;;/m00../s1
SMILES CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@H](NC)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C.CN[C@@H]8[C@H](C)O[C@H](C[C@@H]8OC)O[C@H]9[C@H](C)O[C@H](C[C@@H]9OC)O[C@H]%10[C@@H](C)\C=C\C=C\%11/CO[C@@H]%12[C@H](O)C(=C[C@@H](C(=O)O[C@H]%13C[C@@H](C\C=C\%10/C)O[C@@]%14(C%13)O[C@H](C(C)C)[C@@H](C)C=C%14)[C@]%11%12O)C.OC(=O)c%15ccccc%15.OC(=O)c%16ccccc%16
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 2002.45
Quantity 1 mL
Formula Weight 2001.1056 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.