missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emamectin B1b, TRC
Specifications
Specifications
| Chemical Name or Material | Emamectin B1b |
| CAS | 121424-52-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C48 H73 N O13 |
| Quantity | 2.5 mg |
| Synonym | Emamectin B1b, Avermectin A1a, 5-O-demethyl-25-de(1-methylpropyl)-4''-deoxy-4''-(methylamino)-25-(1-methylethyl)-, (4''R)-, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], avermectin A1a deriv., 4''-Deoxy-4''-epi-methylaminoavermectin B1b, Emamectin B1b |
| InChI Formula | InChI=1S/C48H73NO13/c1-25(2)42-28(5)17-18-47(62-42)23-34-20-33(61-47)16-15-27(4)43(26(3)13-12-14-32-24-55-45-41(50)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-11)44(31(8)57-39)60-38-21-36(53-10)40(49-9)30(7)56-38/h12-15,17-19,25-26,28,30-31,33-45,49 |
| SMILES | CN[C@@H]1[C@H](C)O[C@H](C[C@@H]1OC)O[C@H]2[C@H](C)O[C@H](C[C@@H]2OC)O[C@H]3[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O[C@H]6C[C@@H](C\C=C\3/C)O[C@@]7(C6)O[C@H](C(C)C)[C@@H](C)C=C7)[C@]45O)C |
| Molecular Weight (g/mol) | 872.092 |
| Formula Weight | 871.508 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?