missing translation for 'onlineSavingsMsg'
Learn More

Emamectin B1 Benzoate (Mixture of B1a and B1b), TRC

Product Code. 18043230
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18043230 250 mg 250mg
18032730 2.5 g 2.5g
18033630 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18043230 Supplier Toronto Research Chemicals Supplier No. TRCE520503250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137512-74-4
Molecular Formula C49 H76 N O13 . C7 H5 O2
Molecular Weight (g/mol) 1008.24
SMILES CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H]([NH2+]C)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C.[O-]C(=O)c8ccccc8

Specifications

Quantity 250 mg
Synonym (4' 'R)-4' '-Deoxy-4' '-(methylamino)avermectin B1 Benzoate, Affirm, Denim, Denim (pesticide), Emamectin Benzoate, Emamectin Benzoate (salt), MK 244, Methylamino abamectin Benzoate, Proclaim, Proclaim 05SG, Proclaim 5SG, Revive, Revive (insecticide), Sch 58854, Shot Wan Liquid Formulation, Shot-Wan, Slice
InChI Formula InChI=1S/C49H75NO13.C7H6O2/c1-12-26(2)44-29(5)18-19-48(63-44)24-35-21-34(62-48)17-16-28(4)43(27(3)14-13-15-33-25-56-46-42(51)30(6)20-36(47(52)59-35)49(33,46)53)60-40-23-38(55-11)45(32(8)58-40)61-39-22-37(54-10)41(50-9)31(7)57-39;8-7(9)6-4-2-1-3-5-6/h13-16,18-20,26-27,29,31-32,34-46,50-51,53H,12,17,21-25H2,1-11H3;1-5H,(H,8,9)/b14-13+,28-16+,33-15+;/t26-,27-,29-,31-,32-,34+,35-,36-,37-,38-,39-,40-,41-,42+,43-,44+,45-,46+,48+,49+;/m0./s1
Formula Weight 1007.5606
Chemical Name or Material Emamectin B1 Benzoate (Mixture of B1a and B1b)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.