missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EM 12-FS
GREENER_CHOICE

Product Code. 18764434
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18764434

Brand: Tocris Bioscience™ 7851/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molecular glue; induces degradation of NQAT1

EM12-FS is a selective N-terminal glutamine amidohydrolase 1 (NTAQ1) Degrader. In MOLT4 cells, it downregulates NTAQ1 via cereblon-mediated degradation (IC50 = 256 nM). Requires covalent His353 engagement to recruit and degrade NTAQ1.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[6-[(Fluorosulfonyl)oxy]-1,3-dihydro-1-oxo-2H-isoindol-2-yl]-2,6-piperidinedione
CAS 2839138-44-0
Quantity 50 mg
Target Molecular Glues
Molecular Formula C13H11FN2O6S
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.