missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EM 12-FS
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
EM12-FS is a selective N-terminal glutamine amidohydrolase 1 (NTAQ1) Degrader. In MOLT4 cells, it downregulates NTAQ1 via cereblon-mediated degradation (IC50 = 256 nM). Requires covalent His353 engagement to recruit and degrade NTAQ1.
Specifications
Specifications
| Chemical Name or Material | 3-[6-[(Fluorosulfonyl)oxy]-1,3-dihydro-1-oxo-2H-isoindol-2-yl]-2,6-piperidinedione |
| CAS | 2839138-44-0 |
| Quantity | 10 mg |
| Target | Molecular Glues |
| Molecular Formula | C13H11FN2O6S |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction