missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ELYS Polyclonal antibody specifically detects ELYS in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | ELYS |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AT hook containing transcription factor 1, ELYS, ELYS transcription factor like protein TMBS62, ELYS transcription factor-like protein TMBS62, MST108, MSTP108, protein ELYS, putative AT-hook-containing transcription factor 1, TMBS62 |
| Host Species | Rabbit |
| Immunogen | A syntetic peptide between aa2220 and C-term |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?