missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELYS Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-238CL1
This item is not returnable.
View return policy
Description
ELYS Polyclonal antibody specifically detects ELYS in Human samples. It is validated for Immunoprecipitation,Western Blot,Simple Western
Specifications
| ELYS | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Simple Western | |
| CoraFluor 1 | |
| AT hook containing transcription factor 1, ELYS, ELYS transcription factor like protein TMBS62, ELYS transcription factor-like protein TMBS62, MST108, MSTP108, protein ELYS, putative AT-hook-containing transcription factor 1, TMBS62 | |
| A syntetic peptide between aa2220 and C-term | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 25909 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction