missing translation for 'onlineSavingsMsg'
Learn More

Elvitegravir, Mikromol™

Product Code. 17962776
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize'
100mg
This item is not returnable. View return policy

Product Code. 17962776

missing translation for 'mfr': Mikromol MM3372.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Elvitegravir
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Elvitegravir
CAS 697761-98-1
Analyte or Component Names Elvitegravir
Shipping Condition Room Temperature
Molecular Formula C23 H23 Cl F N O5
Certifications/Compliance ISO 17025
Synonym 3-Quinolinecarboxylic acid, 6-[(3-chloro-2-fluorophenyl)methyl]-1,4-dihydro-1-[(1S)-1-(hydroxymethyl)-2-methylpropyl]-7-methoxy-4-oxo-, 1,4-Dihydro-6-(3-chloro-2-fluorobenzyl)-7-methoxy-1-[(1S)-1-(hydroxymethyl)-2-methylpropyl]-4-oxo-3-quinolinecarboxylic acid, 6-(3-Chloro-2-fluorobenzyl)-1-[(S)-1-hydroxymethyl-2-methylpropyl]-7-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, Elvitegravir, GS 9137, JTK 303
InChI Formula InChI=1S/C23H23ClFNO5/c1-12(2)19(11-27)26-10-16(23(29)30)22(28)15-8-14(20(31-3)9-18(15)26)7-13-5-4-6-17(24)21(13)25/h4-6,8-10,12,19,27H,7,11H2,1-3H3,(H,29,30)/t19-/m1/s1
SMILES COc1cc2N(C=C(C(=O)O)C(=O)c2cc1Cc3cccc(Cl)c3F)[C@H](CO)C(C)C
IUPAC Name 6-[(3-chloro-2-fluorophenyl)methyl]-1-[(2S)-1-hydroxy-3-methylbutan-2-yl]-7-methoxy-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 447.88
Quantity 100 mg
Formula Weight 447.1249 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.