Learn More
Macherey-Nagel™ Elution Buffer N5-EF
Product Code. 30694920
Change view
Click to view available options
Quantity:
3 L
Unit Size:
3 L
Product Code. Quantity unitSize
30694920 3 L 3 L
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30694920 Supplier Macherey-Nagel™ Supplier No. 740796.3
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Elution Buffer N5‐EF to be used with NucleoBond PC EF kits
  • For NucleoBond PC EF kits
Specifications
For Use With (Application) Endotoxin-free plasmid DNA isolation
Quantity 3 L
Solution Type Buffer