missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Elution Buffer for GeneJET Plasmid Midiprep Kit
Shop All Thermo Scientific Products
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
Description
Thermo Scientific GeneJET Elution Buffer is a component of the GeneJET Plasmid Midiprep Kit (Cat. No. K0481) and may be purchased separately. Provided in 60 mL aliquots.
Thermo Scientific GeneJET Elution Buffer is a component of the GeneJET Plasmid Midiprep Kit (Cat. No. K0481) and may be purchased separately. Provided in 60 mL aliquots.
Specifications
Specifications
| Buffer | Elution Buffer |
| For Use With (Application) | Plasmid Prep |
| Product Line | GeneJET |
| Quantity | 60 mL |
| Scale | Midi |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction