missing translation for 'onlineSavingsMsg'
Learn More

Eltrombopag Olamine, MedChemExpress

Product Code. 16456726
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16456726

Brand: Medchem Express HY15306A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eltrombopag Olamine (Eltrombopag diethanolamine salt) is an orally active thrombopoietin receptor nonpeptide agonist.

Eltrombopag Olamine owns thrombopoietic activity, and has been used to research low blood platelet counts with chronic immune thrombocytopenia. Eltrombopag Olamine can be used for the research of cardiovascular. Eltrombopag Olamine also has highly inhibitory effects against multidrug resistant Staphylococcus aureus. Eltrombopag Olamine can induce apoptosis in hepatocellular carcinomab (HCC) as well.

Specifications

Chemical Name or Material Eltrombopag Olamine
CAS 496775-62-3
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C29H36N6O6
Quantity 100 mg
Synonym Eltrombopag diethanolamine salt SB-497115GR
Solubility Information DMSO : ≥ 50 mg/mL (88.55 mM)
SMILES O=C(C1=CC(C2=CC=CC(N/N=C3C(C)=NN(C4=CC=C(C)C(C)=C4)C/3=O)=C2O)=CC=C1)O.NCCO.NCCO
Molecular Weight (g/mol) 564.63
Formula Weight 564.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.