missing translation for 'onlineSavingsMsg'
Learn More

Eltrombopag, MedChemExpress

Product Code. 16486438
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16486438

Brand: Medchem Express HY1530610mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eltrombopag (SB-497115) is an orally active thrombopoietin receptor nonpeptide agonist.

Eltrombopag owns thrombopoietic activity, and has been used to research low blood platelet counts with chronic immune thrombocytopenia. Eltrombopag can be used for the research of cardiovascular. Eltrombopag also has highly inhibitory effects against multidrug resistant Staphylococcus aureus. Eltrombopag can induce apoptosis in hepatocellular carcinomab (HCC) as well.

Specifications

Chemical Name or Material Eltrombopag
CAS 496775-61-2
Color Orange
Purity Grade Notes Research
Molecular Formula C25H22N4O4
Quantity 10 mg
Synonym SB-497115
Solubility Information DMSO : 8.33 mg/mL (18.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC(C2=CC=CC(N/N=C3C(C)=NN(C4=CC=C(C)C(C)=C4)C/3=O)=C2O)=CC=C1)O
Molecular Weight (g/mol) 442.47
Formula Weight 442.47
Percent Purity 98.41%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.