missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eltrombopag, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Eltrombopag owns thrombopoietic activity, and has been used to research low blood platelet counts with chronic immune thrombocytopenia. Eltrombopag can be used for the research of cardiovascular. Eltrombopag also has highly inhibitory effects against multidrug resistant Staphylococcus aureus. Eltrombopag can induce apoptosis in hepatocellular carcinomab (HCC) as well.
Specifications
Specifications
| Chemical Name or Material | Eltrombopag |
| CAS | 496775-61-2 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C25H22N4O4 |
| Quantity | 50 mg |
| Synonym | SB-497115 |
| Solubility Information | DMSO : 8.33 mg/mL (18.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(C1=CC(C2=CC=CC(N/N=C3C(C)=NN(C4=CC=C(C)C(C)=C4)C/3=O)=C2O)=CC=C1)O |
| Molecular Weight (g/mol) | 442.47 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction