missing translation for 'onlineSavingsMsg'
Learn More

Eltrombopag Ethyl Ester, TRC

Product Code. 30386728
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386728 5 mg 5mg
30370342 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386728 Supplier Toronto Research Chemicals Supplier No. TRCE5080105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eltrombopag Ethyl Ester
Analyte or Component Names Eltrombopag Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C27H26N4O4
Synonym 3'-[(2Z)-2-[1-(3,4-Dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy-[1,1'-Biphenyl]-3-carboxylic Acid Ethyl Ester
InChI Formula InChI=1S/C27H26N4O4/c1-5-35-27(34)20-9-6-8-19(15-20)22-10-7-11-23(25(22)32)28-29-24-18(4)30-31(26(24)33)21-13-12-16(2)17(3)14-21/h6-15,30,32H,5H2,1-4H3
Solution Type Neat
SMILES CCOC(=O)c1cccc(c1)c2cccc(N\N=C/3\C(=NN(C3=O)c4ccc(C)c(C)c4)C)c2O
Industry Type Pharmaceutical
Molecular Weight (g/mol) 470.52
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.