missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14203
This item is not returnable.
View return policy
Description
PA1-4203 detects the ETL protein in human samples. PA1-4203 has been successfully used in Western blot procedures. The PA1-4203 immunogen is a synthetic peptide corresponding to residues R(7) S S S N Q D R F I T N D(19) of human ETL.
ETL is a novel seven-transmembrane receptor that belongs to the secretin family of G-protein-coupled peptide hormone receptors and the EGF-TM7 subfamily of receptors. It is the first seven-transmembrane receptor containing EGF-like repeats that is developmentally regulated in the heart.
Specifications
| ELTD1 | |
| Polyclonal | |
| Unconjugated | |
| ADGRL4 | |
| 1110033N21Rik; ADGRL4; Adhesion G protein-coupled receptor L4; EGF, latrophilin and seven transmembrane domain containing 1; EGF, latrophilin and seven transmembrane domain-containing protein 1; EGF, latrophilin seven transmembrane domain containing 1; EGF, latrophilin seven transmembrane domain-containing protein 1; EGF,latrophilin and seven transmembrane domain-containing protein 1; EGF-TM7 receptor; EGF-TM7-latrophilin-related protein; Eltd1; ETL; ETL protein; ETL1; hCG_14667; KPG_003; UNQ202/PRO228 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64123 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9HBW9 | |
| ADGRL4 | |
| Synthetic peptide corresponding to residues R(7) S S S N Q D R F I T N D(19) of human ETL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction