missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELTD1/ETL Antibody (CL4164) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59041AF488
This item is not returnable.
View return policy
Description
ELTD1/ETL Monoclonal antibody specifically detects ELTD1/ETL in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ELTD1/ETL | |
| Monoclonal | |
| Alexa Fluor 488 | |
| EGF, latrophilin and seven transmembrane domain containing 1, EGF, latrophilin and seven transmembrane domain-containing protein 1, ETL protein, ETLEGF-TM7-latrophilin-related protein, KPG_003 | |
| This antibody was developed using a recombinant protein derived from Q9HBW9, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 64123 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4164 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction