missing translation for 'onlineSavingsMsg'
Learn More

ELR510444, MedChemExpress

Product Code. 16466998
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466998

Brand: Medchem Express HY16191

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ELR510444 is a novel microtubule disruptor; inhibits MDA-MB-231 cell proliferation with IC50 of 30.9 nM; not a substrate for the P-glycoprotein drug transporter and retains activity in βIII-tubulin-overexpressing cell lines.

ELR510444 is a novel microtubule disruptor

Specifications

Chemical Name or Material ELR510444
CAS 1233948-35-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H16N2O2S2
Quantity 10 mg
Solubility Information DMSO : ≥ 37 mg/mL (100.42 mM)
SMILES O=S(C1=CC=C(C)C=C1)(NC2=CC(C3=CC=C(C#N)S3)=CC=C2C)=O
Molecular Weight (g/mol) 368.47
Formula Weight 368.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.