missing translation for 'onlineSavingsMsg'
Learn More

Elp4 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18404595 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404595 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404595 Supplier Novus Biologicals Supplier No. NBP212785AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Elp4 Polyclonal antibody specifically detects Elp4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elp4
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C11orf19, chromosome 11 open reading frame 19, dJ68P15A.1, elongation protein 4 homolog (S. cerevisiae), elongator complex protein 4, hELP4, PAX6 neighbor gene protein, PAX6NEB, PAXNEBFLJ20498
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 374 and 424 of human Elongation Protein 4 using the numbering given in entry NP_061913.3 (GeneID 26610).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 26610
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.