missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELP3 Recombinant Rabbit Monoclonal Antibody (JE60-10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544874
This item is not returnable.
View return policy
Description
ELP3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
Specifications
| ELP3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9CZX0, Q9H9T3 | |
| ELP3 | |
| Recombinant protein within human Elp3 aa 1-547/547. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE60-10 | |
| Unconjugated | |
| ELP3 | |
| 2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; elongator complex protein 3; ELP3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100362548, 55140, 74195 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction