missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£164.00 - £442.00
Specifications
| Antigen | ELOVL6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232085
|
Novus Biologicals
NBP3-37909-20ul |
20 μL |
£164.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231118
|
Novus Biologicals
NBP3-37909-100ul |
100 μL |
£442.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELOVL6 Polyclonal antibody specifically detects ELOVL6 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| ELOVL6 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 79071 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast), ELOVL fatty acid elongase 6,3-keto acyl-CoA synthase ELOVL6, FAE, fatty acid elongase 2, long-chain fatty-acyl elongase | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human ELOVL6 (NP_076995.1).,, Sequence:, LMNKRAKFELRKPLVLWSLTLAVFSIFGALRTGAYMVYILMTKGLKQSVCDQGFYNGPVSKFWAYAFVLSKAPELGDTIFIILRKQKLIFLHWYHHITVLL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title